Merck
  • Engineering Escherichia coli coculture systems for the production of biochemical products.
Engineering Escherichia coli coculture systems for the production of biochemical products.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-27)
Haoran Zhang, Brian Pereira, Zhengjun Li, Gregory Stephanopoulos
ABSTRACT
Engineering microbial consortia to express complex biosynthetic pathways efficiently for the production of valuable compounds is a promising approach for metabolic engineering and synthetic biology. Here, we report the design, optimization, and scale-up of an Escherichia coli-E. coli coculture that successfully overcomes fundamental microbial production limitations, such as high-level intermediate secretion and low-efficiency sugar mixture utilization. For the production of the important chemical cis,cis-muconic acid, we show that the coculture approach achieves a production yield of 0.35 g/g from a glucose/xylose mixture, which is significantly higher than reported in previous reports. By efficiently producing another compound, 4-hydroxybenzoic acid, we also demonstrate that the approach is generally applicable for biosynthesis of other important industrial products.
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size