Merck
  • Neutrophil and monocyte toll-like receptor 4, CD11b and reactive oxygen intermediates, and neuroimaging outcomes in preterm infants.
Neutrophil and monocyte toll-like receptor 4, CD11b and reactive oxygen intermediates, and neuroimaging outcomes in preterm infants.
Pediatric research (2015-04-01)
Fiona M O'Hare, William Watson, Amanda O'Neill, Tim Grant, Chike Onwuneme, Veronica Donoghue, Eoghan Mooney, Paul Downey, John Murphy, Anne Twomey, Eleanor J Molloy
ABSTRACT
Activated leukocytes and infection are implicated in neonatal brain injury. Leukocyte surface receptors are increased in stroke models and may be targets for future adjunctive therapies. Serial blood samples were analyzed from preterm infants (n = 51; <32 wk gestation) on days 0, 1, 2, and 7 of life. Monocyte and neutrophil activation were evaluated via flow cytometry at baseline and following endotoxin stimulation ex vivo by measuring CD11b (activation), toll-like receptor 4 (TLR-4; endotoxin recognition) expression, and intracellular reactive oxygen intermediate (ROI) production (function). Control preterm infants with normal neuroimaging had elevated baseline CD11b and TLR-4 expression and ROI production compared with adults as well as a robust immune response following endotoxin stimulation. Preterm infants with abnormal neuroimaging had increased neutrophil TLR-4 and ROI compared with all controls. Preterm infants have a robust immune response compared with adults. Increased TLR-4 expression in preterm infants with abnormal neuroimaging is similar to findings in adult stroke. In addition, ROI production may cause tissue injury. The modulation of these responses may be beneficial in preterm inflammatory disorders.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water