Merck
  • Synthesis and characterization of the antitubercular phenazine lapazine and development of PLGA and PCL nanoparticles for its entrapment.
Synthesis and characterization of the antitubercular phenazine lapazine and development of PLGA and PCL nanoparticles for its entrapment.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
Narjara Silveira, Mariana M Longuinho, Suzana G Leitão, Raphael S F Silva, Maria C Lourenço, Pedro E A Silva, Maria do Carmo F R Pinto, Leida G Abraçado, Priscilla V Finotelli
ABSTRACT
The aim of this work was to develop and characterize nanoparticles as carriers of lapazine, a phenazine derived from β-lapachone; its antimycobacterial activity is described for the first time as a potential treatment for tuberculosis. The lapazine was synthesized, and by using gas chromatography coupled to a flame ionization detector, it was possible to evaluate its purity degree of almost 100%. For better elucidation of the molecular structure, mass spectroscopy and 1H NMR were carried out and compared to the literature values. Lapazine was assayed in vitro against H37Rv Mycobacterium tuberculosis and a rifampicin-resistant strain, with minimum inhibitory concentration values of 3.00 and 1.56 μg mL(-1), respectively. The nanoparticles showed a polydispersity index of 0.16,mean diameter of 188.5 ± 1.7 mm, zeta potential of -15.03 mV, and drug loading of 54.71 mg g(-1) for poly-ε-caprolactone (PCL) nanoparticles and a polydispersity index of 0.318,mean diameter of 197.4 ± 2.7 mm, zeta potential of -13.43 mV and drug loading of 137.07 mg g(-1) for poly(DL-lactide-co-glycolide) (PLGA) nanoparticles. These results indicate that both polymeric formulations have good characteristics as potential lapazine carriers in the treatment of tuberculosis.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%