Merck
  • Annotation of the Staphylococcus aureus Metabolome Using Liquid Chromatography Coupled to High-Resolution Mass Spectrometry and Application to the Study of Methicillin Resistance.
Annotation of the Staphylococcus aureus Metabolome Using Liquid Chromatography Coupled to High-Resolution Mass Spectrometry and Application to the Study of Methicillin Resistance.
Journal of proteome research (2015-10-27)
Sandrine Aros-Calt, Bruno H Muller, Samia Boudah, Céline Ducruix, Gaspard Gervasi, Christophe Junot, François Fenaille
ABSTRACT
Staphylococcus aureus can cause a variety of severe disease patterns and can readily acquire antibiotic resistance; however, the mechanisms by which this commensal becomes a pathogen or develops antibiotic resistance are still poorly understood. Here we asked whether metabolomics can be used to distinguish bacterial strains with different antibiotic susceptibilities. Thus, an efficient and robust method was first thoroughly implemented to measure the intracellular metabolites of S. aureus in an unbiased and reproducible manner. We also placed special emphasis on metabolome coverage and annotation and used both hydrophilic interaction liquid chromatography and pentafluorophenyl-propyl columns coupled to high-resolution mass spectrometry in conjunction with our spectral database developed in-house to identify with high confidence as many meaningful S. aureus metabolites as possible. Overall, we were able to characterize up to 210 metabolites in S. aureus, which represents a substantial ∼50% improvement over previously published data. We then preliminarily compared the metabolic profiles of 10 clinically relevant methicillin-resistant and susceptible strains harvested at different time points during the exponential growth phase (without any antibiotic exposure). Interestingly, the resulting data revealed a distinct behavior of "slow-growing" resistant strains, which show modified levels of several precursors of peptidoglycan and capsular polysaccharide biosynthesis.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%