Merck
  • One-step purification of R-phycoerythrin from the red edible seaweed Grateloupia turuturu.
One-step purification of R-phycoerythrin from the red edible seaweed Grateloupia turuturu.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-06)
Mathilde Munier, Michèle Morançais, Justine Dumay, Pascal Jaouen, Joël Fleurence
ABSTRACT
A one-step chromatographic method for the purification of R-phycoerythrin (R-PE) of Grateloupia turuturu Yamada is described. Native R-PE was obtained with a purity index of 2.89 and a recovery yield of 27% using DEAE-Sepharose Fast Flow chromatography with a three-step increase in ionic strength. The analysis by SDS electrophoresis showed a broad band between 18 and 21kDa in size corresponding to subunits α and β and a low intensity band of 29kDa corresponding to the γ subunit. Two forms of R-PE were identified by gel filtration chromatography: a native form with a molecular weight of 260±5kDa and a dissociated form with a molecular weight of 60±2kDa. The native form presented the characteristic absorption spectrum of R-PE with three absorbance maxima at 498, 540 and 565nm, whereas the dissociated form presented only the 498 and 540nm peaks. Moreover, the two forms displayed two different fluorescence maxima.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O