Merck
  • Assessment of heat shock protein 70 induction by heat in alfalfa varieties and constitutive overexpression in transgenic plants.
Assessment of heat shock protein 70 induction by heat in alfalfa varieties and constitutive overexpression in transgenic plants.
PloS one (2015-05-08)
Nicoletta Ferradini, Rina Iannacone, Stefano Capomaccio, Alessandra Metelli, Nadia Armentano, Lucia Semeraro, Francesco Cellini, Fabio Veronesi, Daniele Rosellini
ABSTRACT
Heat shock proteins (HSPs) are molecular chaperones involved in many cellular functions. It has been shown that mammalian cytosolic HSP70 binds antigenic peptides mediating the activation of the immune system, and that it plays a determining role in tumour immunogenicity. This suggests that HSP70 may be used for the production of conjugated vaccines. Human and plant HSPs share high sequence similarity and some important biological functions in vitro. In addition, plant HSPs have no endotoxic side effects. Extraction of HSP70 from plants for use as vaccine adjuvant requires enhancing its concentration in plant tissues. In this work, we explored the possibility to produce HSP70 in both transgenic and non-transgenic plants, using alfalfa as a model species. First, a transcriptional analysis of a constitutive and an inducible HSP70 genes was conducted in Arabidopsis thaliana. Then the coding sequence of the inducible form was cloned and introduced into alfalfa by Agrobacterium-mediated transformation, and the accumulation of the protein in leaf tissue of transgenic plants was demonstrated. We also tested diverse alfalfa varieties for heat-inducible expression of endogenous HSP70, revealing variety-specific responses to heat shock.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
Glycine
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%