Merck
  • Emmonsiellopsis, a new genus related to the thermally dimorphic fungi of the family Ajellomycetaceae.
Emmonsiellopsis, a new genus related to the thermally dimorphic fungi of the family Ajellomycetaceae.
Mycoses (2015-06-23)
Y Marin-Felix, A M Stchigel, J F Cano-Lira, M Sanchis, E Mayayo, J Guarro
ABSTRACT
Two interesting fungi were isolated from fluvial sediments collected in the North of Spain. They were morphologically related to the thermally dimorphic fungi of the family Ajellomycetaceae, but the analysis of the internal transcribed spacer region of the rDNA, and the domains D1 and D2 of the 28S rRNA gene sequences confirmed that they were different from all the species described in that family. They were accommodated in the new genus Emmonsiellopsis as E. coralliformis sp. nov. and E. terrestris sp. nov. The two species are distinguished mainly by the maximum temper-ature of growth (up to 33 °C for E. coralliformis and to 42 °C for E. terrestris), the dendritic mycelium of E. coralliformis and the conidial ornamentation (verrucose in E. coralliformis and spinulose in E. terrestris). In addition, the phylogenetic data demonstrated that Ajellomyces griseus also represents a new genus within the Ajellomycetaceae, namely Helicocarpus. This new genus is easily distinguished by the lack of asexual morph, the production of brownish gymnothecial ascomata and oblate to lenticular, sparingly pitted ascospores. The proposal of both new genera was confirmed by the analysis of actin gene sequences.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, suitable for electrophoresis
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Pricing and availability is not currently available.
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.