Merck
  • Optimizing photo-mineralization of aqueous methyl orange by nano-ZnO catalyst under simulated natural conditions.
Optimizing photo-mineralization of aqueous methyl orange by nano-ZnO catalyst under simulated natural conditions.
Journal of environmental health science & engineering (2015-05-23)
Ahed Zyoud, Amani Zu'bi, Muath H S Helal, DaeHoon Park, Guy Campet, Hikmat S Hilal
ABSTRACT
Photo-degradation of organic contaminants into non-hazardous mineral compounds is emerging as a strategy to purify water and environment. Tremendous research is being done using direct solar light for these purposes. In this paper we report on optimum conditions for complete mineralization of aqueous methyl orange using lab-prepared ZnO nanopowder catalyst under simulated solar light. Nano-scale ZnO powder was prepared in the lab by standard methods, and then characterized using electronic absorption spectra, photolumenscence emission (PL) spectra, XRD, and SEM. The powder involved a wurtzite structure with ~19 nm particles living in agglomerates. Photo-degradation progressed faster under neutral or slightly acidic conditions which resemble natural waters. Increasing catalyst concentration increased photodegradation rate to a certain limit. Values of catalyst turn over number and degradation percentage increased under higher light intensity, whereas the quantum yield values decreased. The photocatalytic efficiency of nano-ZnO powders in methyl orange photodegradation in water with solar light has been affected by changing the working conditions. More importantly, the process may be used under natural water conditions with pH normally less than 7, with no need to use high concentrations of catalyst or contaminant. The results also highlight the negative impact of possible high concentrations of CO2 on water purification processes. Effects of other added gaseous flows to the reaction mixture are also discussed. ZnO nano-particles are useful catalyst for complete mineralization of organic contaminants in water. Photo-degradation of organic contaminants with ZnO nano-particles, methyl orange being an example, should be considered for future large scale water purification processes under natural conditions.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O