Merck
  • Butyrylated starch intake can prevent red meat-induced O6-methyl-2-deoxyguanosine adducts in human rectal tissue: a randomised clinical trial.
Butyrylated starch intake can prevent red meat-induced O6-methyl-2-deoxyguanosine adducts in human rectal tissue: a randomised clinical trial.
The British journal of nutrition (2015-06-18)
Richard K Le Leu, Jean M Winter, Claus T Christophersen, Graeme P Young, Karen J Humphreys, Ying Hu, Silvia W Gratz, Rosalind B Miller, David L Topping, Anthony R Bird, Michael A Conlon
ABSTRACT
Epidemiological studies have identified increased colorectal cancer (CRC) risk with high red meat (HRM) intakes, whereas dietary fibre intake appears to be protective. In the present study, we examined whether a HRM diet increased rectal O(6)-methyl-2-deoxyguanosine (O(6)MeG) adduct levels in healthy human subjects, and whether butyrylated high-amylose maize starch (HAMSB) was protective. A group of twenty-three individuals consumed 300 g/d of cooked red meat without (HRM diet) or with 40 g/d of HAMSB (HRM+HAMSB diet) over 4-week periods separated by a 4-week washout in a randomised cross-over design. Stool and rectal biopsy samples were collected for biochemical, microbial and immunohistochemical analyses at baseline and at the end of each 4-week intervention period. The HRM diet increased rectal O(6)MeG adducts relative to its baseline by 21% (P < 0.01), whereas the addition of HAMSB to the HRM diet prevented this increase. Epithelial proliferation increased with both the HRM (P < 0.001) and HRM + HAMSB (P < 0.05) diets when compared with their respective baseline levels, but was lower following the HRM + HAMSB diet compared with the HRM diet (P < 0.05). Relative to its baseline, the HRM + HAMSB diet increased the excretion of SCFA by over 20% (P < 0.05) and increased the absolute abundances of the Clostridium coccoides group (P < 0.05), the Clostridium leptum group (P < 0.05), Lactobacillus spp. (P < 0.01), Parabacteroides distasonis (P < 0.001) and Ruminococcus bromii (P < 0.05), but lowered Ruminococcus torques (P < 0.05) and the proportions of Ruminococcus gnavus, Ruminococcus torques and Escherichia coli (P < 0.01). HRM consumption could increase the risk of CRC through increased formation of colorectal epithelial O(6)MeG adducts. HAMSB consumption prevented red meat-induced adduct formation, which may be associated with increased stool SCFA levels and/or changes in the microbiota composition.
Sigma-Aldrich
Heptanoic acid, 96%
Sigma-Aldrich
Heptanoic acid, 97%, FG
Sigma-Aldrich
Heptanoic acid, ≥99.0% (GC)
Sigma-Aldrich
Heptanoic acid, natural, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis