Merck
  • Citrulline does not prevent skeletal muscle wasting or weakness in limb-casted mice.
Citrulline does not prevent skeletal muscle wasting or weakness in limb-casted mice.
The Journal of nutrition (2015-03-06)
Daniel J Ham, Tahnee L Kennedy, Marissa K Caldow, Annabel Chee, Gordon S Lynch, René Koopman
ABSTRACT
Increasing arginine (Arg) availability reduces atrophy in cultured skeletal muscle cells. Supplementation with its metabolic precursor citrulline (Cit) is more effective at improving skeletal muscle Arg concentrations. We tested the hypothesis that Cit supplementation would attenuate skeletal muscle atrophy and loss of function during hindlimb immobilization in mice. Male C57BL/6JArc mice underwent 14 d of unilateral hindlimb immobilization/plaster casting and were supplemented with ~0.81 g Cit · kg⁻¹ · d⁻¹ (CIT group) or Ala (ALA group) mixed into their food. The uncasted contralateral limb (internal control) and an uncasted group (CON) served as controls. Muscle atrophy was evaluated with mass, fiber area, and in situ muscle function. Tibialis anterior (TA) muscle mass [ALA: 37.6 ± 0.92 mg; CIT: 38.3 ± 1.25 mg] and peak tetanic force (ALA: 1150 ± 38.5 mN; CIT: 1150 ± 52.0 mN) were lower (P < 0.001) in the ALA (53.9 ± 0.42 mg) and CIT (1760 ± 28.5 mN) groups than in the CON group. No difference was found between ALA and CIT groups for TA mass, fiber area, or peak force. The mRNA expression of the nitric oxide synthase 2, inducible (Nos2; ~15-fold) and B-cell chronic lymphoid leukemia/lymphoma 2/adenovirus E1B 19 kDa interacting protein 3 (Bnip3; ~17-fold) genes and the ratio of microtubule-associated protein light chain 3BII to 3BI (LC3BII:LC3BI) (50.5% ± 17.7%) were higher (P < 0.05) in the ALA group than in the CON group, suggesting increased autophagy. In the CIT group, Bnip3 mRNA was lower (-70%; P < 0.05) and Nos2 mRNA tended to be lower (-45%; P = 0.05) than in the ALA group, whereas LC3BII:LC3BI was not different from the CON group. Cit treatment of male mice did not affect therapeutically relevant outcome measures such as skeletal muscle mass and peak muscle force after 14 d of hindlimb immobilization.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution