Merck
  • Isolation of three dibenzocyclooctadiene lignans from in vitro cultures of Schisandra chinensis (Turcz.) Baill.--the first report.
Isolation of three dibenzocyclooctadiene lignans from in vitro cultures of Schisandra chinensis (Turcz.) Baill.--the first report.
Die Pharmazie (2015-06-13)
A Szopa, W Kisiel, H Ekiert, A Szewczyk
ABSTRACT
Three dibenzocyclooctadiene lignans: deoxyschizandrin (1), gomisin A (2) and schizandrin (3) were isolated from biomass extracts of Schisandra chinensis (Turcz.) Baill. shoot-differentiating callus cultures. The mentioned lignans were not isolated earlier from in vitro cultures of this plant species. This is the first report concerning on isolation of dibenzocyclooctadiene lignans from in vitro cultures of Schisandra chinensis.
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Rhein