Merck
  • Glutaredoxin 1 (Grx1) Protects Human Retinal Pigment Epithelial Cells From Oxidative Damage by Preventing AKT Glutathionylation.
Glutaredoxin 1 (Grx1) Protects Human Retinal Pigment Epithelial Cells From Oxidative Damage by Preventing AKT Glutathionylation.
Investigative ophthalmology & visual science (2015-03-20)
Xiaobin Liu, Jamieson Jann, Christy Xavier, Hongli Wu
ABSTRACT
Glutaredoxin 1 (Grx1) belongs to the oxidoreductase family and is a component of the endogenous antioxidant defense system. However, its physiological function remains largely unknown. In this study, we investigated whether and how Grx1 overexpression protects the retinal pigment epithelial (RPE) cells against H2O2-induced apoptosis. Human retinal pigment epithelial (ARPE-19) cells were transfected with either a Grx1-containing plasmid or an empty vector. Primary human RPE cells were transfected with Grx1 small interfering RNA (siRNA) or scrambled siRNA. Cell viability was measured with the WST8 assay. Apoptosis was quantitatively measured by annexin V/propidium iodide (PI) double staining. The level of protein glutathionylation (PSSG) was measured by immunoblotting using anti-PSSG antibody. Protein kinase B (AKT) activation was examined by Western blot. Protein kinase B glutathionylation was detected by immunoprecipitation followed by immunoblotting with anti-PSSG antibody. Glutaredoxin 1 overexpression protected ARPE-19 cells from H2O2-induced cell viability loss. Conversely, Grx1 gene knockdown sensitized primary human RPE cells to H2O2. Assessment of apoptosis indicated that cells transfected with the Grx1-containing plasmid were more resistant to H2O2 with fewer cells undergoing apoptosis as compared to empty vector-transfected cells. Hydrogen peroxide-induced PSSG accumulation was also attenuated by Grx1 enrichment. Furthermore, Grx1 overexpression prevented H2O2-induced AKT glutathionylation, resulting in a sustained phospho-AKT elevation in RPE cells. Glutaredoxin 1 can protect RPE cells against oxidative stress-induced apoptosis. The mechanism of this protection is associated with its ability to stimulate the phosphorylation of AKT by preventing AKT glutathionylation. Considering Grx1's protective abilities in RPE cells, Grx1 could be a potential pharmacological target for retinal degenerative diseases.
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, ≥98%, lyophilized powder
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
SAFC
L-Glutathione oxidized
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Glrx
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder