Merck
  • The anion recognition properties of a novel hydrazone based on colorimetric and potentiometric studies.
The anion recognition properties of a novel hydrazone based on colorimetric and potentiometric studies.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
Divya Singhal, Neha Gupta, Ashok Kumar Singh
ABSTRACT
A novel hydrazone, 1,3-bis (2-formylphenoxy) propane bis (2,4-dinitrophenyl hydrazine) (S) has been synthesized and characterized by the NMR, IR, CHNS, UV-Vis, Raman, TEM and XPS analysis. Upon the addition of monohydrogen phosphate (MHP) ion, sudden color change from light yellow to dark violet has occurred. The binding constant of S-MHP was determined by Hill plot. Hydrazone also showed potential response with MHP ion. A number of polymeric membrane electrodes (PME) and coated graphite electrode (CGE) were also fabricated and found that the CGE has revealed better results in comparison to PME in terms of the detection limit of 7.58 × 10(-9) mol L(-1) and Nernstian response.
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
1,3-Dibromopropane, ReagentPlus®, 99%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor