Merck
  • Elevated expression of immunity-related GTPase family M in gastric cancer.
Elevated expression of immunity-related GTPase family M in gastric cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-02-25)
Zongchang Song, Chunliang Guo, Lu Zhu, Pinying Shen, Haitao Wang, Changsheng Guo, Jiahong Tang
ABSTRACT
Recent researches have suggested that autophagy may play critical roles in tumorigenesis. Immunity-related GTPase family M (IRGM) is a human protein highlighted for its contribution to autophagy upon inflammation and infections. Studies have shown that IRGM is involved in the development of several cancers. In the current study, we investigated expression of IRGM and gastric cancer. Levels of messenger RNA (mRNA) and protein were examined by real-time reverse transcription PCR (RT-PCR) and Western blot, respectively. Data showed that mRNA level of IRGM was significantly increased in peripheral blood mononuclear cells (PBMCs) of gastric cancer patients than in PBMCs from healthy controls (p > 0.05). Moreover, both mRNA and protein levels were significantly higher in cancer tissues compared to adjacent noncancerous stomach tissues (1.28-fold, p < 0.001; 1.19-fold, p < 0.01, respectively). However, the level of IRGM seemed not to be affected by Helicobacter pylori infection. In addition, we investigated the correlation between IRGM expression and cancer stages and identified that stage IV patients had upregulated mRNA level and protein level of IRGM in cancer tissues than those stage I patients. Our findings suggest that expression of IRGM is dysregulated in gastric cancer and that the molecule may affect progression of the disease.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri