Merck
  • Hybrid silica monolith for microextraction by packed sorbent to determine drugs from plasma samples by liquid chromatography-tandem mass spectrometry.
Hybrid silica monolith for microextraction by packed sorbent to determine drugs from plasma samples by liquid chromatography-tandem mass spectrometry.
Talanta (2015-06-07)
Israel D de Souza, Diego S Domingues, Maria E C Queiroz
ABSTRACT
The present study (1) reports on the synthesis of two hybrid silica monoliths functionalized with aminopropyl or cyanopropyl groups by the sol-gel process; (2) evaluates these monoliths as selective stationary phase for microextraction by packed sorbent (MEPS) to determine drugs in plasma samples via liquid chromatography-tandem mass spectrometry (LC-MS/MS) in the multiple reactions monitoring (MRM) mode; and (3) discusses important factors related to the optimization of MEPS efficiency as well as the carryover effect. The prepared hybrid silica monoliths consisted of a uniform, porous, and continuous silica monolithic network. The structure of the aminopropyl hybrid silica monolith was more compact than the structure of the cyanopropyl hybrid silica monolith. The Fourier-transform infrared spectroscopy (FTIR) spectra of the hybrid silica monoliths displayed readily identifiable peaks, characteristic of the cyanopropyl and aminopropyl groups. Compared with the aminopropyl hybrid silica phase, the cyanopropyl hybrid silica phase exhibited higher binding capacity for most of the target drugs. The developed method afforded adequate linearity at concentrations ranging from the lower limit of quantification (0.05-1.00 ng mL(-1)) to the upper limit of quantification (40-10,500 ng mL(-1)); the coefficients of determination (r(2)) were higher than 0.9955. The precision of the method presented coefficients of variation (CV) lower than 14%; the relative standard error (RSE) of the accuracy ranged from -12% to 14%. The developed method allowed for simultaneous analysis of five antipsychotics (olanzapine, quetiapine, clozapine, haloperidol, and chlorpromazine) in combination with seven antidepressants (mirtazapine, paroxetine, citalopram, sertraline, imipramine, clomipramine, fluoxetine), two anticonvulsants (carbamazepine and lamotrigine), and two anxiolytics (diazepam and clonazepam) in plasma samples from schizophrenic patients, which should be valuable for therapeutic drug monitoring purposes.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
3-Cyanopropyltriethoxysilane, 98%
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Mirtazapine, ≥98% (HPLC)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dodecylamine, ≥99%
Sigma-Aldrich
Dodecylamine, 98%