Merck
  • The Protective Roles of Selenium on Hepatic Tissue Ultrastructure and Mitochondrial Antioxidant Capacity in Copper-Overloaded Rats.
The Protective Roles of Selenium on Hepatic Tissue Ultrastructure and Mitochondrial Antioxidant Capacity in Copper-Overloaded Rats.
Biological trace element research (2015-03-13)
Rongsheng Su, Huabin Cao, Jiaqiang Pan, Chengmei Li, Yizhou Chen, Zhaoxin Tang
ABSTRACT
The aim of this study was to explore the effects of selenium addition on hepatic tissue ultrastructure and mitochondrial antioxidant capacity in copper-overloaded rats. Eighteen adult male Sprague-Dawley (SD) rats were randomly divided into three groups (n = 6 per group). Each group received 1 mL (intragastrically) of water (control, group I) or water containing copper chloride (CuCl2, 1 mol/L) (group II) or a mixture of CuCl2 (1 mol/L) with sodium selenite (Na2SeO3, 0.05 mol/L) (group III) once daily for 30 days. Histological examination revealed normal hepatocyte structure and no ultrastructural changes in mitochondria in controls. In contrast, group II exhibited severe ultrastructural alterations, fuzzy mitochondrial membranes, irregularly shaped and fragmented rough endoplasmic reticulum (RER), and the high melanin content; group III also exhibited larger amounts of engulfing vesicles (EV) in the cytoplasm. Compared to controls, the CuCl2 treatment lowered (P < 0.05) hepatocyte enzymatic activities and increased lipid peroxidation as indicated by an increase in malondialdehyde (MDA) levels. The effects of CuCl2 were attenuated by simultaneous administration with Na2SeO3. These results indicated that the adverse effects of copper toxicity can be partially attenuated by providing a source of selenium.
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Copper(II) chloride, powder, 99%