Merck
  • Synthesis and Characterization of Hapten-Protein Conjugates for Antibody Production against Cyanogenic Glycosides.
Synthesis and Characterization of Hapten-Protein Conjugates for Antibody Production against Cyanogenic Glycosides.
Journal of food protection (2015-07-22)
Islamiyat Folashade Bolarinwa
ABSTRACT
Consumption of cyanogenic plants can cause serious health problems for humans. The ability to detect and quantify cyanogenic glycosides, capable of generating cyanide, could contribute to prevention of cyanide poisoning from the consumption of improperly processed cyanogenic plants. Hapten-protein conjugates were synthesized with amygdalin and linamarin by using a novel approach. Polyclonal antibodies were generated by immunizing four New Zealand White rabbits with synthesized amygdalin-bovine serum albumin and linamarin-bovine serum albumin immunogen. This is the first time an antibody was produced against linamarin. Antibody titer curves were obtained from all the four rabbits by using a noncompetitive enzyme-linked immunosorbent assay. High antibody titer was obtained at dilutions greater than 1:50,000 from both immunogens. This new method is an important step forward in preventing ingestion of toxic cyanogenic glycosides.
Sigma-Aldrich
α-Hydroxyisobutyronitrile β-D-glucopyranoside, ≥97% (HPLC)
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Potassium carbonate, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
VenPure® SF, powder