Merck
Electric field breakdown in single molecule junctions.
Journal of the American Chemical Society (2015-02-13)
Haixing Li, Timothy A Su, Vivian Zhang, Michael L Steigerwald, Colin Nuckolls, Latha Venkataraman
ABSTRACT
Here we study the stability and rupture of molecular junctions under high voltage bias at the single molecule/single bond level using the scanning tunneling microscope-based break-junction technique. We synthesize carbon-, silicon-, and germanium-based molecular wires terminated by aurophilic linker groups and study how the molecular backbone and linker group affect the probability of voltage-induced junction rupture. First, we find that junctions formed with covalent S-Au bonds are robust under high voltage and their rupture does not demonstrate bias dependence within our bias range. In contrast, junctions formed through donor-acceptor bonds rupture more frequently, and their rupture probability demonstrates a strong bias dependence. Moreover, we find that the junction rupture probability increases significantly above ∼1 V in junctions formed from methylthiol-terminated disilanes and digermanes, indicating a voltage-induced rupture of individual Si-Si and Ge-Ge bonds. Finally, we compare the rupture probabilities of the thiol-terminated silane derivatives containing Si-Si, Si-C, and Si-O bonds and find that Si-C backbones have higher probabilities of sustaining the highest voltage. These results establish a new method for studying electric field breakdown phenomena at the single molecule level.
Sigma-Aldrich
Hexamethylenediamine, technical grade, 70%
Sigma-Aldrich
1,6-Hexanedithiol, 96%
Sigma-Aldrich
1,6-Hexanedithiol, ≥97%, FG
Sigma-Aldrich
Hexamethylenediamine, 98%