Merck
  • Toxicity Evaluation of a New Zn-Doped CuO Nanocomposite With Highly Effective Antibacterial Properties.
Toxicity Evaluation of a New Zn-Doped CuO Nanocomposite With Highly Effective Antibacterial Properties.
Toxicological sciences : an official journal of the Society of Toxicology (2015-03-31)
Paride Mantecca, Elisa Moschini, Patrizia Bonfanti, Umberto Fascio, Ilana Perelshtein, Anat Lipovsky, Giuseppe Chirico, Renato Bacchetta, Luca Del Giacco, Anita Colombo, Aharon Gedanken
ABSTRACT
The increased resistances to conventional antibiotics determine a strong need for new antibacterials, and specific syntheses at the nanoscale promise to be helpful in this field. A novel Zinc-doped Copper oxide nanocomposite (nZn-CuO) has been recently sonochemically synthesized and successfully tested also against multi-drug resistant bacteria. After synthesis and characterization of the physicochemical properties, the new nZn-CuO is here evaluated by the Frog Embyo Teratogenesis Assay-Xenopus test for its toxicological potential and this compared with that of nCuO and nZnO synthesized under the same conditions. No lethal effects are observed, while malformations and growth retardation slightly increase after nZn-CuO exposure. Nevertheless, these effects are smaller than those of nZnO. NP uptake by embryo tissues increase significantly with increasing NP concentrations, while no significant accumulation and adverse effects are seen after exposure to soluble Cu(2+) and Zn(2+) at the concentrations dissolved from the NPs. Key oxidative response genes are upregulated by nZn-CuO, as well as by nCuO and nZnO, suggesting the common mechanism of action. Considering the enhanced biocidal activity shown by the nanocomposite, together with the results presented in this study, we can affirm that the doping of the metal oxide nanoparticles should be considered a useful tool to engineer a safer nano-antibacterial.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
3-Aminobenzoic acid, 98%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis