Merck
  • Comparison of single-stage and two-phase anaerobic sludge digestion systems - Performance and microbial community dynamics.
Comparison of single-stage and two-phase anaerobic sludge digestion systems - Performance and microbial community dynamics.
Chemosphere (2014-08-07)
Yogananda Maspolim, Yan Zhou, Chenghong Guo, Keke Xiao, Wun Jern Ng
ABSTRACT
This study compared reactor performance and the respective microbial community dynamics in the conventional single-stage and 2-phase anaerobic digestion (AD) systems, treating municipal sludge to generate methane. The 2-phase system's COD and VS reduction, and methane production could be maintained throughout the three HRTs tested (p=0.05), which was associated with an increase in organic loading (30d=1.5gCODL(-1)d(-1), 20d=2.2gCODL(-1)d(-1) and 10d=3.5gCODL(-1)d(-1)); but this was not so in the single-stage system where it deteriorated at HRT of 10d (p=0.05) due to impairment of particulate COD reduction. qPCR, DGGE and the subsequent phylogenetic analysis revealed that microbial adaptation occurred as the seed sludge formed a different community in each reactor at 30d HRT; however, no further significant microbial shift occurred at lower HRTs. The presence of specific hydrolytic and acidogenic Flavobacteriales and Clostriales in the acidogenic reactor may have allowed for enhanced hydrolysis and acidogenesis, leading to higher organic loading tolerance at 10d HRT. Methanogenic activity in the acidogenic reactor may have been performed by Methanobacteriales and Methanosarcinaceae. Operation of the acidogenic reactor at neutral pH may have to be considered to ensure the cultivation of propionate oxidising bacteria, which could in turn, prevent reactor "souring" during high load conditions.
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)