Merck
  • Comparison of physicochemical properties of B-type nontraditional starches from different sources.
Comparison of physicochemical properties of B-type nontraditional starches from different sources.
International journal of biological macromolecules (2015-04-15)
Jun Huang, Lingxiao Zhao, Jianmin Man, Juan Wang, Weidong Zhou, Huyin Huai, Cunxu Wei
ABSTRACT
Starches were isolated from rhizomes of Curcuma longa, Canna edulis and Canna indica and bulbs of Lilium lancifolium, and showed a B-type X-ray diffraction pattern. Their physicochemical properties were investigated and compared. These starches showed significantly different granule morphologies and sizes, but all had eccentric hila. The C. longa starch had the lowest content of amylopectin short branch-chain and branching degree and the highest content of amylopectin long branch-chain, and the L. lancifolium starch the highest content of amylopectin short branch-chain and branching degree and the lowest content of amylopectin long branch-chain among the four starches. The L. lancifolium starch had the lowest resistance to gelatinization, and showed the lowest pasting peak, hot and final viscosities, and the C. longa starch had the highest resistance to gelatinization, and showed the highest pasting hot, final and setback viscosities and the lowest pasting breakdown viscosity. The C. longa and L. lancifolium starches possessed very high and low resistance to hydrolysis and digestion, respectively. The above physicochemical properties would be useful for the applications of B-type starches in food and nonfood industries.
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%