Merck
  • Preparation and thermo-reversible gelling properties of protein isolate from defatted Antarctic krill (Euphausia superba) byproducts.
Preparation and thermo-reversible gelling properties of protein isolate from defatted Antarctic krill (Euphausia superba) byproducts.
Food chemistry (2015-06-05)
Yanchao Wang, Ruo Wang, Yaoguang Chang, Ying Gao, Zhaojie Li, Changhu Xue
ABSTRACT
Protein isolate was prepared from defatted Antarctic krill (Euphausia superba) byproducts by 0.1 M NaOH extraction. Maximum yield of krill protein isolate reached 28.66% by precipitation at pH 4.6. Krill protein isolate demonstrated its excellent nutritional values through amino acid composition and in vitro digestibility. Thermal transition of krill protein isolate was determined by differential scanning calorimetry measurement. Extrapolated values of glass transition temperature (Tg) and denaturation temperature (Td) of krill protein isolate were 33.8 °C and 80.3 °C when the heating rate was 2 °C/min. Dispersions of krill protein isolate generated self-supported gels at concentrations above 100 g/L without the addition of salt or other additives. A noticeable enhancement of gel strength was induced through cooling. Gels with krill protein isolate displayed a thermo-reversible behavior under repeating heating/cooling cycles, which was primarily due to the formation and disruption of hydrogen bonds. Network strength of protein gels was strongly dependent on protein concentrations.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein