Merck
Evaluation of ice-tea quality by DART-TOF/MS.
Journal of mass spectrometry : JMS (2015-10-28)
Aleš Rajchl, Jana Prchalová, Vojtěch Kružík, Rudolf Ševčík, Helena Čížková
ABSTRACT
DART (Direct Analysis in Real Time) coupled with Time-of-Flight Mass Spectrometry (TOF/MS) has been used for analyses of ice-teas. The article focuses on quality and authenticity of ice-teas as one of the most important tea-based products on the market. Twenty-one samples of ice-teas (black and green) were analysed. Selected compounds of ice-teas were determined: theobromine, caffeine, total phenolic compounds, total soluble solids, total amino acid concentration, preservatives and saccharides were determined. Fingerprints of DART-TOF/MS spectra were used for comprehensive assessment of the ice-tea samples. The DART-TOF/MS method was used for monitoring the following compounds: citric acid, caffeine, saccharides, artificial sweeteners (saccharin, acesulphame K), and preservatives (sorbic and benzoic acid), phosphoric acid and phenolic compounds. The measured data were subjected to a principal components analysis. The HPLC and DART-TOF/MS methods were compared in terms of determination of selected compounds (caffeine, benzoic acid, sorbic acid and saccharides) in the ice-teas. The DART-TOF/MS technique seems to be a suitable method for fast screening, testing quality and authenticity of tea-based products.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Potassium Sorbate, analytical standard
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium sorbate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology