Merck
  • Transforming berberine into its intestine-absorbable form by the gut microbiota.
Transforming berberine into its intestine-absorbable form by the gut microbiota.
Scientific reports (2015-07-16)
Ru Feng, Jia-Wen Shou, Zhen-Xiong Zhao, Chi-Yu He, Chao Ma, Min Huang, Jie Fu, Xiang-Shan Tan, Xiao-Yang Li, Bao-Ying Wen, Xi Chen, Xin-Yi Yang, Gang Ren, Yuan Lin, Yangchao Chen, Xue-Fu You, Yan Wang, Jian-Dong Jiang
ABSTRACT
The gut microbiota is important in the pathogenesis of energy-metabolism related diseases. We focused on the interaction between intestinal bacteria and orally administered chemical drugs. Oral administration of berberine (BBR) effectively treats patients with metabolic disorders. However, because BBR exhibits poor solubility, its absorption mechanism remains unknown. Here, we show that the gut microbiota converts BBR into its absorbable form of dihydroberberine (dhBBR), which has an intestinal absorption rate 5-fold that of BBR in animals. The reduction of BBR to dhBBR was performed by nitroreductases of the gut microbiota. DhBBR was unstable in solution and reverted to BBR in intestine tissues via oxidization. Heat inactivation of intestinal homogenate did not inhibit dhBBR oxidization, suggesting the process a non-enzymatic reaction. The diminution of intestinal bacteria via orally treating KK-Ay mice with antibiotics decreased the BBR-to-dhBBR conversion and blood BBR; accordingly, the lipid- and glucose-lowering efficacy of BBR was reduced. Conclusively, the gut microbiota reduces BBR into its absorbable form of dhBBR, which then oxidizes back to BBR after absorption in intestine tissues and enters the blood. Thus, interaction(s) between the gut microbiota and orally administrated drugs may modify the structure and function of chemicals and be important in drug investigation.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%
Sigma-Aldrich
[Bis(trifluoroacetoxy)iodo]benzene, 97%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid