Merck
  • Development and evaluation of gastroretentive raft forming systems incorporating curcumin-Eudragit® EPO solid dispersions for gastric ulcer treatment.
Development and evaluation of gastroretentive raft forming systems incorporating curcumin-Eudragit® EPO solid dispersions for gastric ulcer treatment.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-06)
Nattha Kerdsakundee, Sirima Mahattanadul, Ruedeekorn Wiwattanapatapee
ABSTRACT
Novel raft forming systems incorporating curcumin-Eudragit® EPO solid dispersions were developed to prolong the gastric residence time and provide for a controlled release therapy of curcumin to treat gastric ulcers. The solid dispersions of curcumin with Eudragit® EPO were prepared by the solvent evaporation method at various ratios to improve the solubility and the dissolution of curcumin. The optimum weight ratio of 1:5 for curcumin to Eudragit® EPO was used to incorporate into the raft forming systems. The raft forming formulations were composed of curcumin-Eudragit® EPO solid dispersions, sodium alginate as a gelling polymer and calcium carbonate for generating divalent Ca(2+) ions and carbon dioxide to form a floating raft. All formulations formed a gelled raft in 1min and sustained buoyancy on the 0.1N hydrochloric acid (pH 1.2) surface with a 60-85% release of curcumin within 8h. The curative effect on the acetic acid-induced chronic gastric ulcer in rats was determined. The curcumin raft forming formulations at 40mg/kg once daily showed a superior curative effect on the gastric ulcer in terms of the ulcer index and healing index than the standard antisecretory agent: lansoprazole (1mg/kg, twice daily) and a curcumin suspension (40mg/kg, twice daily). These studies demonstrated that the new raft forming systems containing curcumin solid dispersions are promising carriers for a stomach-specific delivery of poorly soluble lipophilic compounds.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis