Merck
Toxicity of oral cadmium intake: Impact on gut immunity.
Toxicology letters (2015-06-09)
Marina Ninkov, Aleksandra Popov Aleksandrov, Jelena Demenesku, Ivana Mirkov, Dina Mileusnic, Anja Petrovic, Ilijana Grigorov, Lidija Zolotarevski, Maja Tolinacki, Dragan Kataranovski, Ilija Brceski, Milena Kataranovski
ABSTRACT
Gastrointestinal tract is one of the main targets of cadmium (Cd), an important food and drinking water contaminant. In the present study, the effect of subchronic (30 days) oral (in water) intake of 5ppm and 50ppm of cadmium on immune responses in the gut was examined in rats. Cadmium consumption resulted in reduction of bacteria corresponding to Lactobacillus strain, tissue damage and intestinal inflammation [increases in high mobility group box 1 (HMGB1 molecules), superoxide dismutase (SOD) and catalase (CAT) activity and proinflammatory cytokine (TNF, IL-1β, IFN-γ, IL-17) content]. Draining (mesenteric) lymph node (MLN) stress response was observed [elevation of MLN glutathione (GSH) and metallothionein (MT) mRNA levels] and stimulation of both adaptive [cellularity, proliferation, proinflammatory (IFN-γ and IL-17) MLN cell cytokine responses] as well as innate immune activity (increases in numbers of NK and CD68(+) cells, oxidative activities, IL-1β). In contrast to proinflammatory milieu in MLN, decreased or unchanged antiinflammatory IL-10 response was observed. Stimulation of immune activities of MLN cells have, most probably, resulted from sensing of cadmium-induced tissue injury, but also from bacterial antigens that breached compromised intestinal barrier. These effects of cadmium should be taken into account when assessing dietary cadmium as health risk factor.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Cadmium chloride
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%