Merck
  • Antimicrobial susceptibility of Listeria monocytogenes isolates from human cases in northern Italy, 2008-2010: MIC determination according to EUCAST broth microdilution method.
Antimicrobial susceptibility of Listeria monocytogenes isolates from human cases in northern Italy, 2008-2010: MIC determination according to EUCAST broth microdilution method.
Journal of chemotherapy (Florence, Italy) (2014-04-12)
Moira Madeo, Rosario Musumeci, Anna Maria Laura Careddu, Ettore Amato, Mirella Maria Pontello, Clementina Elvezia Cocuzza
ABSTRACT
Susceptibility of 96 Listeria monocytogenes human isolates collected in northern Italy between 2008 and 2010, to 15 antimicrobials, was investigated. Minimum inhibitory concentration (MIC) was evaluated by means of the standardized broth microdilution method, according to the European Committee on Antimicrobial Susceptibility Testing (EUCAST) international guidelines. All L. monocytogenes human isolates were susceptible to penicillin G (MIC(90)≤0·06 μg/ml), meropenem (MIC(90)≤0·06 μg/ml), and erythromycin (MIC(90) = 0·12 μg/ml). Susceptibility to the other tested antimicrobials could not be interpreted due to the lack of breakpoint values although two (2%) isolates were shown to have tetracycline MICs above EUCAST epidemiological cut-off values (ECOFF). Bactericidal activity for amoxicillin, gentamicin, and levofloxacin was generally observed at concentrations 2-4 times higher than MIC values. Though L. monocytogenes human strains, isolated in the north of Italy, appear to be susceptible to most antimicrobial agents used in human therapy, this study provides new data for epidemiological surveillance and clinical breakpoints definition.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
Azithromycin