Merck
  • Retinoid resistance and multifaceted impairment of retinoic acid synthesis in glioblastoma.
Retinoid resistance and multifaceted impairment of retinoic acid synthesis in glioblastoma.
Glia (2015-05-07)
Benito Campos, Sarah Weisang, Florian Osswald, Ramadan Ali, Georg Sedlmeier, Josephine Bageritz, Jan-Philipp Mallm, Christian Hartmann, Andreas von Deimling, Odillia Popanda, Violaine Goidts, Christoph Plass, Andreas Unterberg, Peter Schmezer, Jürgen Burhenne, Christel Herold-Mende
ABSTRACT
Measuring concentrations of the differentiation-promoting hormone retinoic acid (RA) in glioblastoma tissues would help to understand the reason why RA treatment has been inefficient in clinical trials involving brain tumor patients. Here, we apply a recently established extraction and measurement protocol to screen glioblastoma tissues for the levels of the RA precursor retinol and biologically active RA. Combining this approach with mRNA analyses of 26 tumors and 8 normal brains, we identify a multifaceted disturbance of RA synthesis in glioblastoma, involving multiple aldehyde dehydrogenase 1 family and retinol dehydrogenase enzymes. Through database studies and methylation analyses, we narrow down chromosomal deletions and aberrant promoter hypermethylation as potential mechanisms accounting for these alterations. Employing chromatin immunoprecipitation analyses and cell-culture studies, we further show that chromatin at RA target genes is poised to RA substitution, but most glioblastoma cell cultures are completely resistant to RA treatment. This paradoxical RA response is unrelated to alternative RA signaling through the fatty acid-binding protein 5/peroxisome proliferator-activated receptor delta axis. Our data suggest a multifaceted disturbance of RA synthesis in glioblastoma and contribute to reconsider current RA treatment strategies.
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)