Merck
  • Amorphastilbol exerts beneficial effects on glucose and lipid metabolism in mice consuming a high-fat-diet.
Amorphastilbol exerts beneficial effects on glucose and lipid metabolism in mice consuming a high-fat-diet.
International journal of molecular medicine (2015-06-03)
Woojung Lee, Jungyeob Ham, Hak Cheol Kwon, Goo Yoon, Gyu-Un Bae, Yong Kee Kim, Su-Nam Kim
ABSTRACT
In the present study, the anti-diabetic effects of amorphastilbol (APH) from Amorpha fruticosa (AF) were evaluated in high-fat-diet (HFD) mice. HFD-induced blood glucose and insulin levels are significantly reduced in AF extract or APH treatment groups. HFD-induced weight gain was reduced by AF treatment, which is accompanied by reduction of fat mass and adipocyte size and number in white adipose tissues. Furthermore, total cholesterol and low-density lipoprotein-cholesterol levels are decreased in AF- or APH-treated mice. In addition, AF and APH are able to improve insulin sensitivity through inhibition of protein tyrosine phosphatase 1B, a negative regulator of the insulin-signaling pathway. Taken together, the data suggest that AF has beneficial effects on glucose and lipid metabolism and its pharmacological effects are driven, in part, by its active component, APH. Therefore, AF and APH can be used as potential therapeutic agents against type 2 diabetes and associated metabolic disorders, including obesity, by enhancing glucose and lipid metabolism.
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))