Merck
  • Factor-inhibiting HIF-1 (FIH-1) is required for human vascular endothelial cell survival.
Factor-inhibiting HIF-1 (FIH-1) is required for human vascular endothelial cell survival.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-04-04)
Serafim Kiriakidis, Anne-Theres Henze, Ilona Kruszynska-Ziaja, Konstantinos Skobridis, Vassiliki Theodorou, Ewa M Paleolog, Massimiliano Mazzone
ABSTRACT
Factor-inhibiting hypoxia-inducible factor (HIF)-1 (FIH-1) is an asparaginyl β-hydroxylase enzyme that was initially found to hydroxylate the HIF-α, preventing its transcriptional activity and leading to adaptive responses to hypoxia. More recently, other substrates, such as neurogenic locus notch homolog (Notch), have been found to be alternative FIH targets, but the biologic relevance of this regulation was never investigated. Given the key function of Notch in angiogenesis, we here investigate the role of FIH/Notch signaling in endothelial cells. We report that FIH-1 silencing in HUVECs results in reduced growth and increased apoptosis. The knockdown of FIH is associated with increased Notch2 activity, leading to enhanced expression of the Notch target hairy/enhancer-of-split related with YRPW motif protein 1 (Hey-1). Consistent with recent findings showing that Notch2 suppresses survivin (a key inhibitor of apoptosis), FIH targeting in HUVECs leads to selective repression of survivin in endothelial cells, thus promoting cell apoptosis and growth arrest. Our data support the concept that FIH-1 may interact with Notch2 and repress its activity, thereby playing a critical role in controlling the survival of vascular endothelial cells. These findings might pave the way toward novel, antiangiogenic strategies in disorders that are characterized by excessive vascular growth, such as cancer and rheumatoid arthritis.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
BIS-TRIS
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG