Merck
  • Model-based description of peptide retention on doped reversed-phase media.
Model-based description of peptide retention on doped reversed-phase media.
Journal of chromatography. A (2015-07-08)
Rushd Khalaf, Nicola Forrer, Gianluca Buffolino, Alessandro Butté, Massimo Morbidelli
ABSTRACT
Reversed-phase (RP) chromatography is one of the main tools for the preparative purification of therapeutic peptides. In previous works [1,2], a new type of RP chromatography, doped reversed-phase chromatography (DRP) was presented. By adding small amounts (up to 15% of the surface ligands) of repulsive ion exchange ligands to a traditional RP material, significant improvements in peptide purification performance were observed, at the same or in similar operating conditions. These improvements included increases in selectivity in diluted conditions (up to twice as high), increases in yield in preparative conditions (up to 20% higher) and in productivity in preparative conditions (up to twice as high), when compared to RP materials [2]. A proper physical model is developed in this work to quantitatively explain and rationalize this behavior. The developed model is then used to correctly fit the retention data of several peptides in different buffering conditions. The increase in selectivity is related to a controlled decrease in free surface area available for adsorption due to the ionic ligands creating a repulsive sphere the analytes cannot enter. This decrease in adsorption surface is calculated using Debye-Hückel theory, and in combination with linear solvent strength theory, allows for the quantitative description of peptide retention on DRP media.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%