Merck
Engineering the ovarian cycle using in vitro follicle culture.
Human reproduction (Oxford, England) (2015-03-19)
Robin M Skory, Yuanming Xu, Lonnie D Shea, Teresa K Woodruff
ABSTRACT
Can cultured follicles model the ovarian cycle, including follicular- and luteal-phase hormone synthesis patterns and ovulation? Under gonadotrophin stimulation, murine follicles grown in an encapsulated three-dimensional system ovulate in vitro and murine and human follicle hormone synthesis mimics follicular and luteal phases expected in vivo. Studies of the human ovary and follicle function are limited by the availability of human tissue and lack of in vitro models. We developed an encapsulated in vitro follicle growth (eIVFG) culture system, which preserves 3D follicular structure. Thus far, the alginate system has supported the culture of follicles from mice, dog, rhesus macaque, baboon and human. These studies have shown that cultured follicles synthesize steroid hormones similar to those observed during the follicular phase in vivo. Cultured murine follicles were treated with human chorionic gonadotrophin (hCG) and epidermal growth factor (EGF) and either assayed for luteinization or removed from alginate beads and assayed for ovulation. Human follicles were also cultured, treated with follicle-stimulating hormone (FSH), hCG and EGF to mimic gonadotrophin changes throughout the ovarian cycle, and culture medium was assayed for hormone production. Murine and human follicles were cultured in alginate hydrogel and hormone production [17β-estradiol, progesterone, inhibin A, inhibin B, activin A and anti-Müllerian hormone (AMH)] was quantified in medium by enzyme-linked immuno assay (ELISA). Human ovarian tissue was acquired from females between 6 and 34 years of age with a cancer diagnosis. These participants were undergoing ovarian tissue cryopreservation at National Physicians Cooperative sites as part of the Oncofertility Consortium. When grown in this system, 96% of mouse follicles ovulated in response to hCG and released meiotically competent eggs. Ovulated follicles recapitulated transcriptional, morphologic and hormone synthesis patterns post-luteinizing hormone (LH/hCG). In addition to rodent follicles, individual human follicles secreted steroid and peptide hormones that mimicked the patterns of serum hormones observed during the menstrual cycle. This was a descriptive study of an in vitro model of ovulation and the ovarian hormone cycle. The ovulation studies were limited to murine tissue and further studies are needed to optimize conditions using other species. The eIVFG system reliably phenocopies the in vivo ovarian cycle and provides a new tool to study human follicle biology and the influence of cycling female hormones on other tissue systems in vitro. This work was supported by NIH U54 HD041857, NIH U54 HD076188, NIH UH2 E5022920, NIH UH3 TR001207 and F30 AG040916 (R.M.S.). The content is solely the responsibility of the authors and does not necessarily represent the official views of the NIH. The funders had no role in study design, data collection and analysis, decision to publish or preparation of the manuscript.
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
SB−3CT, ≥98% (HPLC), powder
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride