Merck
  • Behavior of POP-calcium carbonate hydrogel as bone substitute with controlled release capability: a study in rat.
Behavior of POP-calcium carbonate hydrogel as bone substitute with controlled release capability: a study in rat.
Journal of biomedical materials research. Part A (2015-03-27)
Anne Handrini Dewi, Ika Dewi Ana, Joop Wolke, John Jansen
ABSTRACT
Gypsum or calcium sulfate (CS) or plaster of Paris (POP) is considered as a fast degradable material that usually resorbs before the bone defect area is completely filled by new bone. In this study, the incorporation of CaCO3 hydrogel into POP in different compositions was proposed to enhance the bone biological activity of POP and to decrease its degradability. The mechanical and degradation properties of the various materials were characterized by in vitro analysis. Subsequently, the materials were inserted into cylindrically sized bone defects as created into the femoral condyle of rats and left in situ for 1, 4, and 8 weeks. Histological analysis of the retrieved specimens indicated that the addition of CaCO3 hydrogel into POP increased bone formation, angiogenesis and collagen density and resulted into faster bone formation and maturation. It was also confirmed that the degradation rate of the POP decreased by the addition of CaCO3 hydrogel. The in vivo findings did corroborate with the in vitro analysis. In conclusion, the incorporation of CaCO3 hydrogel provides a promising technology to improve the properties of POP, the oldest biomaterial used for bone grafting.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%