Merck
  • Mechanical stretch activates mammalian target of rapamycin and AMP-activated protein kinase pathways in skeletal muscle cells.
Mechanical stretch activates mammalian target of rapamycin and AMP-activated protein kinase pathways in skeletal muscle cells.
Molecular and cellular biochemistry (2015-05-15)
Naoya Nakai, Fuminori Kawano, Ken Nakata
ABSTRACT
Cellular protein synthesis is believed to be antagonistically regulated by mammalian target of rapamycin (mTOR) and AMP-activated protein kinase (AMPK) signaling pathways. In the present study, we examined the relationship between mTOR/p70 S6 kinase (p70S6K) and AMPK in response to mechanical stretch. C2C12 myoblasts were grown on a silicone elastomer chamber to confluence and further cultured in differentiation medium for 4 days to form multinucleated myotubes. Cells were subjected to 15% cyclic uniaxial stretch for 4 h at a frequency of 1 Hz. Phosphorylation of p70S6K at threonine 389 and AMPK at threonine 172 of the catalytic α subunit were concomitantly increased by mechanical stretch. Stimulation of the mTOR pathway by adding leucine and insulin increased the phosphorylation of p70S6K without inactivation of AMPK. In contrast, addition of compound C, a pharmacological inhibitor of AMPK, increased the phosphorylation of p70S6K in stretched cells. Activation of AMPK by the addition of 5-amino-4-imidazolecarboxamide ribonucleoside reduced the phosphorylation of p70S6K in response to mechanical stretch. In conclusion, crosstalk between mTOR and AMPK signaling was not tightly regulated in response to physiological stimuli, such as mechanical stress and/or nutrients. However, pharmacological modulation of AMPK influenced the mTOR/p70S6K signaling pathway.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)