Merck
  • Comparative Studies on Polyphenolic Composition, Antioxidant and Diuretic Effects of Nigella sativa L. (Black Cumin) and Nigella damascena L. (Lady-in-a-Mist) Seeds.
Comparative Studies on Polyphenolic Composition, Antioxidant and Diuretic Effects of Nigella sativa L. (Black Cumin) and Nigella damascena L. (Lady-in-a-Mist) Seeds.
Molecules (Basel, Switzerland) (2015-05-29)
Claudia-Crina Toma, Neli-Kinga Olah, Laurian Vlase, Cristina Mogoșan, Andrei Mocan
ABSTRACT
This study was performed to evaluate the phenolic profile, antioxidant and diuretic effects of black cumin and lady-in-a-mist seeds. In the phenolic profile, differences between the two species are significant. Qualitative and quantitative analyses of the phenolic compounds were performed using a HPLC-UV/MS method. Hyperoside was the only identified flavonoid glycoside (1.08 ± 0.01 μg∙g-1 dw plant material), in the N. damascena extract. Regarding the flavonol profile, kaempferol was identified before the hydrolysis, only in the N. sativa extract (6.06 ± 0.02 μg∙g-1 dw plant material) and quercetin only in N. damascena seeds (14.35 ± 0.02 μg∙g-1 dw plant material). The antioxidant potential of the two species was tested through several electron transfer assays, which indicated, excepting for the FRAP assay, N. damascena as exhibiting a higher free radical scavenging activity. The diuretic activity of the two extracts was tested using a rat-experimental model on acute diuresis. Administration of the ethanolic extract of N. sativa (100 mg∙kg-1) resulted in a significant increase in urine volume, although less than found with the reference drug; in addition N. damascena extract did not present a diuretic effect. In reference to the elimination of Na+, K+ and uric acid, the black cumin extract exhibited a higher natriuretic than kaluretic effect and a similar uricosuric effect with control and N. damascena. For N. damascena, the Na+/K+ ratio was sub unitary, but not due to an increasing of the kaluretic effect, but mostly to a decrease of Na+ excretion.
Sigma-Aldrich
Caftaric acid, ≥97.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Aluminum chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis