Merck
  • Dissipative solitons and backfiring in the electrooxidation of CO on Pt.
Dissipative solitons and backfiring in the electrooxidation of CO on Pt.
Scientific reports (2015-11-11)
Philipp R Bauer, Antoine Bonnefont, Katharina Krischer
ABSTRACT
Collisions of excitation pulses in dissipative systems lead usually to their annihilation. In this paper, we report electrochemical experiments exhibiting more complex pulse interaction with collision survival and pulse splitting, phenomena that have rarely been observed experimentally and are only poorly understood theoretically. Using spatially resolved in-situ Fourier transform infrared spectroscopy (FTIR) in the attenuated total reflection configuration, we monitored reaction pulses during the electrochemical oxidation of CO on Pt thin film electrodes in a flow cell. The system forms quasi-1d pulses that align parallel to the flow and propagate perpendicular to it. The pulses split once in a while, generating a second solitary wave in the backward moving direction. Upon collision, the waves penetrate each other in a soliton-like manner. These unusual pulse dynamics could be reproduced with a 3-component reaction-diffusion-migration model with two inhibitor species, one of them exhibiting a long-range spatial coupling. The simulations shed light on existence criteria of such dissipative solitons.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size