Merck
  • Nutritional values and bioactive components of under-utilised vegetables consumed by indigenous people in Malaysia.
Nutritional values and bioactive components of under-utilised vegetables consumed by indigenous people in Malaysia.
Journal of the science of food and agriculture (2014-11-21)
Norsuhaila Abdul Wahab, Rohaina Ahdan, Zabidah Ahmad Aufa, Kin Weng Kong, Mohd Hafizan Johar, Zalilah Shariff Mohd, Amin Ismail
ABSTRACT
Diverse plants species in the forest remain under-utilised and they are mainly consumed only by local people. However, increasing issues in food security prompted the present study, which explores the nutritional and antioxidant aspects of Malaysian under-utilised vegetables. The studied vegetables were Paku Nyai (Stenochlaena palustris), Cemperai (Champereia manillana), Maman Pasir (Cleome viscose), Dudung (Erechtites valerianifolia) and Semambuk (Ardisia pendula). Overall, these vegetables exhibited a low proximal content but they were high in vitamin C [7.07-1263 mg kg(-1) edible fresh sample (EFS)] and β-carotene content (18.4-43.9 mg kg(-1) kg(-1) EFS). Cemperai had the highest calcium content (565 mg kg(-1) EFS), whereas Semambuk had the highest total phenolic content [28.21 g gallic acid equivalents kg(-1) edible dried sample (EDS)] and antioxidant activity (86.1%) measured using β-carotene bleaching assay. Maman Pasir contained the highest total flavonoid content (39.99 g CE kg(-1) EDS) and 1,1-diphenyl-2-picryl hydrazyl radical scavenging activity (82.2%). The extracts of these vegetables had significantly prevented the oxidation of haemoglobin and low-density lipoprotein, which yielded a reduced production of malondialdehyde. Semambuk and Maman Pasir are potent to be used as new food and functional food sources as they are rich in nutrients and antioxidants.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)