Merck
  • Psoralen Inactivation of Viruses: A Process for the Safe Manipulation of Viral Antigen and Nucleic Acid.
Psoralen Inactivation of Viruses: A Process for the Safe Manipulation of Viral Antigen and Nucleic Acid.
Viruses (2015-11-17)
Katherine Schneider, Loni Wronka-Edwards, Melissa Leggett-Embrey, Eric Walker, Peifang Sun, Brian Ondov, Travis H Wyman, M J Rosovitz, Sherry S Bohn, James Burans, Tadeusz Kochel
ABSTRACT
High consequence human pathogenic viruses must be handled at biosafety level 2, 3 or 4 and must be rendered non-infectious before they can be utilized for molecular or immunological applications at lower biosafety levels. Here we evaluate psoralen-inactivated Arena-, Bunya-, Corona-, Filo-, Flavi- and Orthomyxoviruses for their suitability as antigen in immunological processes and as template for reverse transcription PCR and sequencing. The method of virus inactivation using a psoralen molecule appears to have broad applicability to RNA viruses and to leave both the particle and RNA of the treated virus intact, while rendering the virus non-infectious.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)