Merck
  • Paralcaligenes ginsengisoli sp. nov., isolated from ginseng cultivated soil.
Paralcaligenes ginsengisoli sp. nov., isolated from ginseng cultivated soil.
Antonie van Leeuwenhoek (2015-07-01)
Jong-Pyo Kang, Ngoc-Lan Nguyen, Yeon-Ju Kim, Van-An Hoang, Kwi-Sik Bae, Deok-Chun Yang
ABSTRACT
A novel bacterial strain DCY104(T) isolated from soil of a ginseng field in Yeoncheon County, Republic of Korea is described in this study. Cells were short rod-shaped, motile by mean of one polar flagellum, strictly aerobic, Gramreaction negative, oxidase and catalase-positive. 16S rRNA gene sequence analysis showed that strain DCY104(T) shared highest similarity 98.2 % to Paralcaligenes ureilyticus GR24-5(T), and from 97.7 to 97.1 % with other type strains belong to the genera Candidimonas, Pusillimonas and Parapusillimonas Otherwise, phylogenetic trees analyses indicated that strain DCY104(T) belongs to a single group with P. ureilyticus GR24-5(T) that was distinct to other genera. The major polar lipids were phosphatidylmonomethylethanolamine, phosphatidylethanolamine, phosphatidylglycerol, and diphosphatidylglycerol. The major cellular fatty acids consisted of C16:0, cyclo-C17:0, and summed feature 8 (comprising C18:1 ω7c and/or C18:1 ω6c). The predominant polyamine was putrescine. The ubiquinone was Q-8. The genomic DNA G+C content was 55.9 mol%. These data in combination with the presence of one polar flagellum and positive activity of urease confirmed the placement of strain DCY104(T) in the genus Paralcaligenes. DNA-DNA relatedness between strain DCY104(T) and P. ureilyticus KACC 13888(T) was 40 %. The differences in the profiles of polar lipids, fatty acids and phenotypic characteristics in combination with DNA-DNA relatedness delineated strain DCY104(T) and P. ureilyticus KACC 13888(T). In summary, taxonomic analyses in this study demonstrated that strain DCY104(T) represents a novel species within the genus Paralcaligenes, for which we propose the name Paralcaligenes ginsengisoli. The type strain is DCY104(T) (= KCTC 42406(T) = JCM 30746(T)).
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
1,8-Diaminooctane, 98%
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)