Merck
  • Identification of Anti-Long Chain Saturated Fatty Acid IgG Antibodies in Serum of Patients with Type 2 Diabetes.
Identification of Anti-Long Chain Saturated Fatty Acid IgG Antibodies in Serum of Patients with Type 2 Diabetes.
Mediators of inflammation (2015-12-04)
Dequina A Nicholas, Lorena M Salto, Ava M Boston, Nan Sun Kim, Marco Larios, W Lawrence Beeson, Anthony F Firek, Carlos A Casiano, William H R Langridge, Zaida Cordero-MacIntyre, Marino De Leon
ABSTRACT
High levels of serum long chain saturated fatty acids (LCSFAs) have been associated with inflammation in type 2 diabetes. Dietary SFAs can promote inflammation, the secretion of IgG antibodies, and secretion of the proinflammatory cytokine IL-1β. This study characterizes anti-LCSFA IgG antibodies from patients with type 2 diabetes. Serum samples from several cohorts with type 2 diabetes were analyzed for the presence of anti-LCSFA IgG, the cytokine IL-1β, and nonesterified fatty acids. Anti-LCSFA IgG was isolated from patient samples and used for in vitro characterization of avidity and specificity. A cohort participating in En Balance, a diabetes health education program that improved diabetes management, tested positive for anti-LCSFA IgG. Following the 3-month program, the cohort showed a significant reduction in anti-LCSFA IgG levels. Anti-LCSFA antibodies isolated from these patients demonstrated high avidity, were specific for long chain SFAs, and correlated with serum fatty acids in patients with managed type 2 diabetes. Interestingly, anti-LCSFA IgG neutralized PA-induced IL-1β secretion by dendritic cells. Our data shows that nonesterified SFAs are recognized by IgG antibodies present in human blood. The identification of anti-LCSFA IgG antibodies in human sera establishes a basis for further exploration of lipid induced immune responses in diabetic patients.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ethyl chloroformate, 97%