Merck
  • Action of anti-M₃muscarinic acetylcholine receptor IgG of primary Sjögren's syndrome on the enzymatic antioxidant system in rat submandibular gland.
Action of anti-M₃muscarinic acetylcholine receptor IgG of primary Sjögren's syndrome on the enzymatic antioxidant system in rat submandibular gland.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2015-03-03)
Silvia Reina, Marcelo Rodríguez, Graciela Stranieri, Enri Borda
ABSTRACT
We demonstrate that serum immunoglobulin G (IgG) directed against glandular M3 muscarinic acetylcholine receptors (M₃mAChR) and pilocarpine triggers the increment of superoxide dismutase (SOD) and catalase (CAT) and the production of nitric oxide (NO) and prostaglandin E₂(PGE₂). Enzyme-linked immunosorbent assay (ELISA) was performed in the presence of the human M₂mAChR synthetic peptide as antigen to detect in serum of pSS patients the autoantibodies. Further, SOD and CAT specific activity and NO were determined chemically in the presence of anti-M₃mAChR IgG and pilocarpine. The level of PGE₂generation in the presence of autoantibody and pilocarpine was determined by ELISA. An association between anti-M₂mAChR autoantibodies and pilocarpine given the increment of the specific activity of SOD and CAT in the serum of pSS patients and in the rat submandibular gland was observed. As a result of this action, M₃synthetic peptide and atropine abrogated the stimulatory action. The L-type calcium channel, calcium/calmodulin complex and COX-2 inhibitors selectively blocked the increment of the specific activity of SOD and CAT in the rat submandibular gland. An increased production of NO and PGE₂by the cholinergic autoantibody and pilocarpine was also detected. On the basis of these results, the increment of the specific activity of SOD and CAT in pSS patients as compared to control healthy individuals may be seen as a defensive reaction to the increment of the amount of ROS, which becoming uncontrollable, leads to irreversible cellular and tissue damage.
Sigma-Aldrich
Sodium carbonate, powder, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Tri(2-furyl)phosphine, 99%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O