Merck
  • Designing greener plasticizers: Effects of alkyl chain length and branching on the biodegradation of maleate based plasticizers.
Designing greener plasticizers: Effects of alkyl chain length and branching on the biodegradation of maleate based plasticizers.
Chemosphere (2015-04-29)
Hanno C Erythropel, Tobin Brown, Milan Maric, Jim A Nicell, David G Cooper, Richard L Leask
ABSTRACT
The ubiquitous presence of the plasticizer di (2-ethylhexyl) phthalate (DEHP) in the environment is of concern due to negative biological effects associated with it and its metabolites. In particular, the metabolite mono (2-ethylhexyl) phthalate (MEHP) is a potential endocrine disruptor. Earlier work had identified the diester di (2-ethylhexyl) maleate (DEHM) as a potential greener candidate plasticizer to replace DEHP, yet its biodegradation rate was reported to be slow. In this study, we modified the side chains of maleate diesters to be linear (i.e., unbranched) alkyl chains that varied in length from ethyl to n-octyl. The plasticization efficiency of these compounds blended into PVC at 29 wt.% increased with the overall length of the molecule, but all compounds performed as well as or better than comparable samples with DEHP. Tests conducted with the equally long DEHM and dihexyl maleate (DHM) showed that branching has no effect on glass transition temperature (Tg) reduction efficiency. Biodegradation experiments with the common soil bacterium Rhodococcus rhodocrous in the presence of the plasticizer showed acceptable hydrolysis rates of maleates with unbranched side chains, while the branched DEHM showed almost no degradation. The addition of hexadecane as auxiliary carbon source improved hydrolysis rates. Temporary buildup of the respective monoester of the compounds were observed, but only in the case of the longest molecule, dioctyl maleate (DOM), did this buildup lead to growth inhibition of the bacteria. Maleates with linear side chains, if designed and tested properly, show promise as potential candidate plasticizers as replacements for DEHP.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Diethyl maleate, ≥96%, FG
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Maleic anhydride, 99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Diethyl maleate, 97%
Sigma-Aldrich
Bis(2-ethylhexyl) maleate, 90%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Formaldehyde diethyl acetal, absolute, over molecular sieve (H2O ≤0.01%), ≥99.0% (GC)
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)
Sigma-Aldrich
Pentadecane, ≥99%
Sigma-Aldrich
Dibutyl maleate, 96%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG