Merck
  • Identification of Cell Surface Straight Chain Poly-N-Acetyl-Lactosamine Bearing Protein Ligands for VH4-34-Encoded Natural IgM Antibodies.
Identification of Cell Surface Straight Chain Poly-N-Acetyl-Lactosamine Bearing Protein Ligands for VH4-34-Encoded Natural IgM Antibodies.
Journal of immunology (Baltimore, Md. : 1950) (2015-10-28)
Neelima M Bhat, Christopher M Adams, Yi Chen, Marcia M Bieber, Nelson N H Teng
ABSTRACT
B cell binding and cytotoxicity by human VH4-34-encoded Abs of the IgM isotype has been well documented. A VH4-34-IgM has recently shown a favorable early response in a phase 1 trial for treatment of B cell acute lymphoblastic leukemia. Although its B cell ligand has been identified as straight chain poly-N-acetyl-lactosamine (SC-PNAL), the carrier of the sugar moiety has not been identified. Using nanoelectrospray ionization mass spectrometry, we identify the metabolic activation related protein complex of CD147-CD98 as a major carrier of poly-N-acetyl-lactosamine (SC-PNAL) on human pre-B cell line Nalm-6. Previous studies have suggested CD45 as the SC-PNAL carrier for VH4-34-encoded IgG Abs. Because Nalm-6 is CD45 negative, human peripheral blood B lymphocytes and human B cell line, Reh, with high CD45 expression, were examined for SC-PNAL carrier proteins. Western blot analysis shows that the CD147-98 complex is indeed immunoprecipitated by VH4-34-encoded IgMs from human peripheral blood B lymphocytes and human B cell lines, Reh, OCI-Ly8, and Nalm-6. However, CD45 is immunoprecipitated only from peripheral B lymphocytes, but not from Reh despite the high expression of CD45. These results suggest that human B cells retain SC-PNAL on the CD147-98 complex, but modulate the sugar moiety on CD45. Because the carbohydrate moiety may act as a selecting Ag for VH4-34 autoantibody repertoire, its differential expression on proteins may provide a clue to the intricate atypical regulation of the VH4-34 gene.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Cholera Toxin B subunit, FITC conjugate, lyophilized powder
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)