Merck
  • The use of dried blood spots for quantification of 15 antipsychotics and 7 metabolites with ultra-high performance liquid chromatography - tandem mass spectrometry.
The use of dried blood spots for quantification of 15 antipsychotics and 7 metabolites with ultra-high performance liquid chromatography - tandem mass spectrometry.
Drug testing and analysis (2014-08-19)
Lisbeth Patteet, Kristof E Maudens, Christophe P Stove, Willy E Lambert, Manuel Morrens, Bernard Sabbe, Hugo Neels
ABSTRACT
Therapeutic drug monitoring of antipsychotics is important in optimizing individual therapy. In psychiatric populations, classical venous blood sampling is experienced as frightening. Interest in alternative techniques, like dried blood spots (DBS), has consequently increased. A fast and easy to perform DBS method for quantification of 16 antipsychotics (amisulpride, aripiprazole, asenapine, bromperidol, clozapine, haloperidol, iloperidone, levosulpiride, lurasidone, olanzapine, paliperidone, pipamperone, quetiapine, risperidone, sertindole and zuclopenthixol) and 8 metabolites was developed. DBS were prepared using 25 μL of whole blood and extraction of complete spots was performed using methanol: methyl-t-butyl-ether (4:1). After evaporation, the extract was reconstituted in the mobile phase and 10 μL were injected on an ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). Separation using a C18 column and gradient elution with a flow rate of 0.5 mL/min resulted in a 6-min run-time. Ionization was performed in positive mode and a dynamic MRM method was applied. Median recovery was 66.4 % (range 28.7-84.5%). Accuracy was within the acceptance criteria, except for pipamperone (LLOQ and low concentration) and lurasidone (low concentration). Imprecision was only aberrant for lurasidone at low and medium concentration. All compounds were stable during 1 month at room temperature, 4 °C and -18 °C. Lurasidone was unstable when the extract was stored for 12 h on the autosampler. Absolute matrix effects (ME) (median 66.1%) were compensated by the use of deuterated IS (median 98.8%). The DBS method was successfully applied on 25-μL capillary DBS from patients and proved to be a reliable alternative for quantification of all antipsychotics except for olanzapine and N-desmethylolanzapine.
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
(S)-(−)-Sulpiride, ≥98% (titration)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Sigma-Aldrich
Pipamperone dihydrochloride, ~99% (HPLC), powder
Sigma-Aldrich
Paliperidone, ≥98% (HPLC)
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Clozapine
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Aripiprazole, ≥98% (HPLC)
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis