Merck
  • Digoxin ameliorates autoimmune arthritis via suppression of Th17 differentiation.
Digoxin ameliorates autoimmune arthritis via suppression of Th17 differentiation.
International immunopharmacology (2015-03-31)
Jennifer Lee, Seungye Baek, Jaeseon Lee, Juhyun Lee, Dong-Gun Lee, Mi-Kyung Park, Mi-La Cho, Sung-Hwan Park, Seung-Ki Kwok
ABSTRACT
Digoxin is a cardiac glycoside that is commonly used to treat heart failure. Based on its known anti-inflammatory effect, this study was undertaken to investigate the effect of digoxin on collagen-induced arthritis (CIA) and to delineate the underlying mechanism. Digoxin or vehicle was injected intraperitoneally thrice weekly in mice with CIA, from day 7 or day 35 after immunization to investigate preventive or therapeutic effect, respectively. The incidence and severity of arthritis was evaluated. Digoxin treatment suppressed the incidence of arthritis and joint inflammation in mice with CIA. The expression of IL-17 and other proinflammatory cytokines, including IL-1β, IL-6, TNF-α and IL-21, were markedly reduced in the arthritic joints of digoxin-treated CIA mice. Th17 cells and CD4(+) pSTAT3(+) cells were less frequently observed in the spleen of digoxin-treated CIA mice than controls. The mRNA expression of IL-17 and ROR γt was consistently lower in total splenocytes or draining lymph node cells obtained from digoxin-treated CIA mice. Digoxin also reduced in vitro Th17 differentiation and LPS-stimulated IgG production. The number of osteoclasts in the arthritic joint was lower in digoxin-treated mice, whereas digoxin treatment did not directly suppress in vitro osteoclastogenesis. Our findings suggest that digoxin can regulate Th17 and reciprocally promote Treg cells and suppress joint inflammation and bone erosion in CIA. Digoxin may be a therapeutic option by targeting pathogenic Th17 and immunoglobulin production, for treatment of autoimmune arthritis and other Th17-related diseases.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%