Merck
  • New insights into the human brain proteome: Protein expression profiling of deep brain stimulation target areas.
New insights into the human brain proteome: Protein expression profiling of deep brain stimulation target areas.
Journal of proteomics (2015-04-08)
Joaquín Fernández-Irigoyen, María Victoria Zelaya, Estela Perez-Valderrama, Enrique Santamaría
ABSTRACT
Deep brain stimulation (DBS) is a neurosurgical procedure that provides therapeutic benefits for movement and affective disorders. The nucleus basalis of Meynert (NBM) and substantia nigra (SN) are considered target areas to apply DBS. Even though the degeneration of NBM and SN underlies the cognitive decline observed in neurological diseases, the protein knowledge derived from both areas is scarce. We have characterized the proteome present in both subcortical brain areas using the Triple TOF 5600 mass spectrometer, identifying 2775 and 3469 proteoforms in NBM and SN respectively. Data mining of MS-generated proteomic data have revealed that: i) 675 proteins tend to localize to synaptic ending, ii) 61% of the global dataset is also present in human CSF and/or plasma, and iii) 894 proteins have not been previously identified in healthy brain by MS. The correlation of NBM and SN proteomic expression profiles with human brain transcriptome data available at Allen Brain Atlas has revealed protein evidence for 250 genes considered with brain-wide expression and 112 genes with region-specific expression in human brain. In addition, protein datasets have been classified according to their chromosomal origin, increasing the current proteome coverage in healthy human brain. The nucleus basalis of Meynert and substantia nigra are brain areas of clinical interest to apply the deep brain stimulation (DBS) technology in neurosurgery. Our proteomic characterization has revealed 675 proteins involved in the regulation of synaptic transmission, electrical machinery, and neurotransmitter release in both DBS target areas. Moreover, 2599 identified proteins present capacity to be secreted to the CSF and plasma. Our data contribute to a further step towards the characterization of the anatomical atlas of the human brain proteome, detecting 652 proteins that are common between different basal ganglia structures. This article is part of a Special Issue entitled: HUPO 2014.
Product Description
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG