Merck
  • Iron Oxide and Titanium Dioxide Nanoparticle Effects on Plant Performance and Root Associated Microbes.
Iron Oxide and Titanium Dioxide Nanoparticle Effects on Plant Performance and Root Associated Microbes.
International journal of molecular sciences (2015-10-09)
David J Burke, Nicole Pietrasiak, Shu F Situ, Eric C Abenojar, Mya Porche, Pawel Kraj, Yutthana Lakliang, Anna Cristina S Samia
ABSTRACT
In this study, we investigated the effect of positively and negatively charged Fe₃O₄ and TiO₂ nanoparticles (NPs) on the growth of soybean plants (Glycine max.) and their root associated soil microbes. Soybean plants were grown in a greenhouse for six weeks after application of different amounts of NPs, and plant growth and nutrient content were examined. Roots were analyzed for colonization by arbuscular mycorrhizal (AM) fungi and nodule-forming nitrogen fixing bacteria using DNA-based techniques. We found that plant growth was significantly lower with the application of TiO₂ as compared to Fe₃O₄ NPs. The leaf carbon was also marginally significant lower in plants treated with TiO₂ NPs; however, leaf phosphorus was reduced in plants treated with Fe₃O₄. We found no effects of NP type, concentration, or charge on the community structure of either rhizobia or AM fungi colonizing plant roots. However, the charge of the Fe₃O₄ NPs affected both colonization of the root system by rhizobia as well as leaf phosphorus content. Our results indicate that the type of NP can affect plant growth and nutrient content in an agriculturally important crop species, and that the charge of these particles influences the colonization of the root system by nitrogen-fixing bacteria.
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C