Merck
  • Different immune cells mediate mechanical pain hypersensitivity in male and female mice.
Different immune cells mediate mechanical pain hypersensitivity in male and female mice.
Nature neuroscience (2015-06-30)
Robert E Sorge, Josiane C S Mapplebeck, Sarah Rosen, Simon Beggs, Sarah Taves, Jessica K Alexander, Loren J Martin, Jean-Sebastien Austin, Susana G Sotocinal, Di Chen, Mu Yang, Xiang Qun Shi, Hao Huang, Nicolas J Pillon, Philip J Bilan, YuShan Tu, Amira Klip, Ru-Rong Ji, Ji Zhang, Michael W Salter, Jeffrey S Mogil
ABSTRACT
A large and rapidly increasing body of evidence indicates that microglia-to-neuron signaling is essential for chronic pain hypersensitivity. Using multiple approaches, we found that microglia are not required for mechanical pain hypersensitivity in female mice; female mice achieved similar levels of pain hypersensitivity using adaptive immune cells, likely T lymphocytes. This sexual dimorphism suggests that male mice cannot be used as proxies for females in pain research.
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse