Merck
  • Expression of O(6)-methylguanine-DNA methyltransferase causes lomustine resistance in canine lymphoma cells.
Expression of O(6)-methylguanine-DNA methyltransferase causes lomustine resistance in canine lymphoma cells.
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire (2015-07-02)
Satoshi Kambayashi, Kouji Minami, Yuka Ogawa, Takehiro Hamaji, Chung Chew Hwang, Masaya Igase, Hiroko Hiraoka, Takako Shimokawa Miyama, Shunsuke Noguchi, Kenji Baba, Takuya Mizuno, Masaru Okuda
ABSTRACT
The DNA repair protein O (6)-methylguanine-DNA methyltransferase (MGMT) causes resistance to nitrosoureas in various human cancers. In this study, we analyzed the correlation between canine lymphomas and MGMT in vitro. Two of five canine lymphoma cell lines required higher concentrations of lomustine to inhibit cell growth by 50%, but their sensitivity to the drug increased when they were cultured with an MGMT inhibitor. Fluorometric oligonucleotide assay and real-time polymerase chain reaction of these cell lines revealed MGMT activity and high MGMT mRNA expression, respectively. We analyzed the methylation status of the CpG islands of the canine MGMT gene by the bisulfite-sequencing method. Unlike human cells, the canine lymphoma cell lines did not show significant correlation between methylation status and MGMT suppression levels. Our results suggest that in canine lymphoma MGMT activity may influence sensitivity to nitrosoureas; thus, inhibition of MGMT activity would benefit nitrosourea-resistant patients. Additional studies are necessary to elucidate the mechanism of regulation of MGMT expression.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%