Merck
  • Effect of Leflunomide on the Abnormal Expression of Lipid Rafts and F-Actin in B Lymphocytes from Patients with Systemic Lupus Erythematosus.
Effect of Leflunomide on the Abnormal Expression of Lipid Rafts and F-Actin in B Lymphocytes from Patients with Systemic Lupus Erythematosus.
Journal of immunology research (2015-06-20)
Guang Fu Dong, Xiao Zhang, De Ning He, Ling Li, Guang Feng Zhang
ABSTRACT
To investigate the possible changes in B cell subsets and in B cell expression patterns of lipid rafts (LRs) and F-actin in patients with SLE and whether leflunomide treatment may have effect on these changes. The B cell subsets and LRs expression were determined by flow cytometry and confocal microscopy, and F-actin expression was examined by confocal microscopy. CD27(+)IgD(+) B cell subsets were significantly decreased while CD38(+)CD95(+) B cell subsets increased in SLE patients. The LRs levels of B cells were remarkably increased and positively correlated with SLEDAI and anti-dsDNA titer in SLE patients. The expression level of LRs was significantly higher in CD38(+) B cells than CD38(-) B cells and negatively correlated with C3 levels. The increased expression of LRs was associated with reduced expression of F-actin in the B cells from active SLE patients. Furthermore, in vitro treatment of the cells with A771726 reduced the expression level of LRs, attenuated the overaggregation of LRs, and normalized the distribution of F-actin. There were abnormalities in B cell subsets and LRs and F-actin expression of B cell from SLE patients. Modulation of B cell expression of LRs and F-actin by LEF could be a potential therapeutic target for SLE.
Sigma-Aldrich
CTB, ≥98% (HPLC)